Learn More
CPC Scientific 5FAM-Ala-Ala-Glu-Glu-Ile-Tyr-Ala-Ala-Arg-Arg-Gly-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110247
Encompass
ONE-LETTER SEQUENCE: 5FAM-AAEEIYAARRG-NH2MOLECULAR FORMULA: C72H94N18O22MOLECULAR WEIGHT:1563.7STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: Chan, Perry M., H. Peter Nestler, and W. Todd Miller. "Investigating the substrate specificity of the HER2/Neu tyrosine kinase using peptide libraries." Cancer Letters 160, no. 2 (2000): 159-169.
Catalog No. 50-211-0247 Supplier CPC Scientific Supplier No. PROK016A
ONE-LETTER SEQUENCE: 5FAM-AAEEIYAARRG-NH2MOLECULAR FORMULA: C72H94N18O22MOLECULAR WEIGHT:1563.7STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: Chan, Perry M., H. Peter Nestler, and W. Todd Miller. "Investigating the substrate specificity of the HER2/Neu tyrosine kinase using peptide libraries." Cancer Letters 160, no. 2 (2000): 159-169.