Learn More
CPC Scientific 5FAM-Ala-Glu-Glu-Ile-Tyr-Gly-Val-Leu-Ala-Lys-Lys-Lys-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110239
Encompass
ONE-LETTER SEQUENCE: 5FAM-AEEIYGVLAKKK-NH2MOLECULAR FORMULA: C83H116N16O23MOLECULAR WEIGHT:1706STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0239 Supplier CPC Scientific Supplier No. PROK012A
ONE-LETTER SEQUENCE: 5FAM-AEEIYGVLAKKK-NH2MOLECULAR FORMULA: C83H116N16O23MOLECULAR WEIGHT:1706STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases