Learn More
CPC Scientific 5FAM-Ala-Lys-Glu-Leu-Asp-Gln-Gly-Ser-Leu-Cys-Thr-Ser-Phe-Val-Gly-Thr-Leu-Gln-NH2 (acetate salt) 0.5MG
SDP
Catalog No. 502110261
Encompass
ONE-LETTER SEQUENCE: 5FAM-AKELDQGSLCTSFVGTLQ-NH2MOLECULAR FORMULA: C102H144N22O34S1MOLECULAR WEIGHT:2254.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0261 Supplier CPC Scientific Supplier No. PROK023A
ONE-LETTER SEQUENCE: 5FAM-AKELDQGSLCTSFVGTLQ-NH2MOLECULAR FORMULA: C102H144N22O34S1MOLECULAR WEIGHT:2254.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases