Learn More
CPC Scientific 5FAM-Ala-Lys-Arg-Arg-Arg-Leu-Ser(PO3H2)-Ser(PO3H2)-Leu-Arg-Ala-OH 1MG
SDP
Catalog No. 502110284
Encompass
SEQUENCE: 5FAM-Ala-Lys-Arg-Arg-Arg-Leu-Ser(PO3H2)-Ser(PO3H2)-Leu-Arg-Ala-OHONE-LETTER SEQUENCE: 5FAM-AKRRRL-Ser(PO3H2)-Ser(PO3H2)-LRAMOLECULAR FORMULA: C75H116N24O26P2MOLECULAR WEIGHT: 1831.9STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES:
SKU(s): PROK-035A, PROK-035B
Catalog No. 50-211-0284 Supplier CPC Scientific Supplier No. PROK035B
SEQUENCE: 5FAM-Ala-Lys-Arg-Arg-Arg-Leu-Ser(PO3H2)-Ser(PO3H2)-Leu-Arg-Ala-OHONE-LETTER SEQUENCE: 5FAM-AKRRRL-Ser(PO3H2)-Ser(PO3H2)-LRAMOLECULAR FORMULA: C75H116N24O26P2MOLECULAR WEIGHT: 1831.9STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES:
SKU(s): PROK-035A, PROK-035B