Learn More
CPC Scientific 5FAM-Ala-Leu-Lys-Leu-Val-Arg-Tyr-Pro-Ser-Phe-Val-Ile-Thr-Ala-Lys-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110281
Encompass
ONE-LETTER SEQUENCE: 5FAM-ALKLVRYPSFVITAK-NH2MOLECULAR FORMULA: C103H147N21O24MOLECULAR WEIGHT:2063.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0281 Supplier CPC Scientific Supplier No. PROK034A
ONE-LETTER SEQUENCE: 5FAM-ALKLVRYPSFVITAK-NH2MOLECULAR FORMULA: C103H147N21O24MOLECULAR WEIGHT:2063.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases