Learn More
Aat Bioquest 5-FAM Alkyne
SDP
Catalog No. 502979190
Encompass_Preferred
5-FAM alkyne is an excellent building block for developing fluoresceinated bioconjugates by reacting with an azide group.
Catalog No. 50-297-9190 Supplier Aat Bioquest Supplier No. 132
5-FAM alkyne is an excellent building block for developing fluoresceinated bioconjugates by reacting with an azide group.