Learn More
CPC Scientific 5FAM-Ala-Thr-Gly-Pro-Leu-Ser-Pro-Gly-Pro-Phe-Gly-Arg-Arg-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110255
Encompass
ONE-LETTER SEQUENCE: 5FAM-ATGPLSPGPFGRRMOLECULAR FORMULA: C79H103N19O22MOLECULAR WEIGHT:1670.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0255 Supplier CPC Scientific Supplier No. PROK020A
ONE-LETTER SEQUENCE: 5FAM-ATGPLSPGPFGRRMOLECULAR FORMULA: C79H103N19O22MOLECULAR WEIGHT:1670.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases