Learn More
Aat Bioquest 5-FAM Azide
SDP
Catalog No. 502979170
Encompass_Preferred
5-FAM azide is an excellent building block for developing fluoresceinated bioconjugates by reacting with an alkyne group.
Catalog No. 50-297-9170 Supplier Aat Bioquest Supplier No. 131
5-FAM azide is an excellent building block for developing fluoresceinated bioconjugates by reacting with an alkyne group.