Learn More
Aat Bioquest 5-FAM cadaverine
SDP
Catalog No. 502979219
Encompass_Preferred
FAM cadaverine is an excellent building block for developing fluoresceinated bioconjugates.
Catalog No. 50-297-9219 Supplier Aat Bioquest Supplier No. 128
FAM cadaverine is an excellent building block for developing fluoresceinated bioconjugates.