Learn More
CPC Scientific 5FAM-Asp-Glu-Glu-Ile-Tyr-Glu-Glu-Leu-Lys-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110268
Encompass
ONE-LETTER SEQUENCE: 5FAM-DEEIYEELK-NH2MOLECULAR FORMULA: C72H89N11O26MOLECULAR WEIGHT:1524.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0268 Supplier CPC Scientific Supplier No. PROK026B
ONE-LETTER SEQUENCE: 5FAM-DEEIYEELK-NH2MOLECULAR FORMULA: C72H89N11O26MOLECULAR WEIGHT:1524.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases