Learn More
CPC Scientific 5FAM-Glu-Arg-Met-Arg-Pro-Arg-Lys-Arg-Gln-Gly-Ser-Val-Arg-Arg-Arg-Val-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110286
Encompass
ONE-LETTER SEQUENCE: 5FAM-ERMRPRKRQGSVRRRV-NH2MOLECULAR FORMULA: C104H166N40O26S1MOLECULAR WEIGHT:2424.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0286 Supplier CPC Scientific Supplier No. PROK036B
ONE-LETTER SEQUENCE: 5FAM-ERMRPRKRQGSVRRRV-NH2MOLECULAR FORMULA: C104H166N40O26S1MOLECULAR WEIGHT:2424.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases