Learn More
CPC Scientific 5FAM-Glu-Arg-Arg-Lys-Ser-Lys-Ser-Gly-Ala-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110500
Encompass
ONE-LETTER SEQUENCE: 5FAM-ERRKSKSGA-NH2MOLECULAR FORMULA: C61H86N18O19MOLECULAR WEIGHT:1375.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0500 Supplier CPC Scientific Supplier No. SUBS030A
ONE-LETTER SEQUENCE: 5FAM-ERRKSKSGA-NH2MOLECULAR FORMULA: C61H86N18O19MOLECULAR WEIGHT:1375.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases