Learn More
CPC Scientific 5FAM-Gly-Glu-Glu-Ile-Tyr-Gly-Glu-Phe-Asp-NH2 1MG
SDP
Catalog No. 502110288
Encompass
SEQUENCE: 5FAM-Gly-Glu-Glu-Ile-Tyr-Gly-Glu-Phe-Asp-NH2ONE-LETTER SEQUENCE: 5FAM-GEEIYGEFD-NH2MOLECULAR FORMULA: C68H74N10O24MOLECULAR WEIGHT: 1415.4STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES:
SKU(s): PROK-037A, PROK-037B
Catalog No. 50-211-0288 Supplier CPC Scientific Supplier No. PROK037B
SEQUENCE: 5FAM-Gly-Glu-Glu-Ile-Tyr-Gly-Glu-Phe-Asp-NH2ONE-LETTER SEQUENCE: 5FAM-GEEIYGEFD-NH2MOLECULAR FORMULA: C68H74N10O24MOLECULAR WEIGHT: 1415.4STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES:
SKU(s): PROK-037A, PROK-037B