Learn More
CPC Scientific 5FAM-Gly-Glu-Ile-Leu-Ser-Arg-Arg-Pro-Ser-Tyr-Arg-Lys-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110290
Encompass
ONE-LETTER SEQUENCE: 5FAM-GEILSRRPSYRK-NH2MOLECULAR FORMULA: C84H119N23O23MOLECULAR WEIGHT:1819STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0290 Supplier CPC Scientific Supplier No. PROK038B
ONE-LETTER SEQUENCE: 5FAM-GEILSRRPSYRK-NH2MOLECULAR FORMULA: C84H119N23O23MOLECULAR WEIGHT:1819STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases