Learn More
CPC Scientific 5FAM-Gly-Glu-Tyr-Ala-Ala-Glu-Tyr-Ala-Asp-Asp-Ala-Tyr-Ala-Ala-Asp-Tyr-Tyr-Ala-Ala-NH2 1MG
SDP
Catalog No. 502110291
Encompass
ONE-LETTER SEQUENCE: 5FAM-GEYAAEYADDAYAADYYAA-NH2MOLECULAR FORMULA: C114H130N20O40MOLECULAR WEIGHT:2420.4STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0291 Supplier CPC Scientific Supplier No. PROK039A
ONE-LETTER SEQUENCE: 5FAM-GEYAAEYADDAYAADYYAA-NH2MOLECULAR FORMULA: C114H130N20O40MOLECULAR WEIGHT:2420.4STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases