Learn More
CPC Scientific 5FAM-Gly-Gly-Gly-Pro-Ala-Thr(PO3H2)-Pro-Lys-Lys-Ala-Lys-Lys-Leu-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110293
Encompass
ONE-LETTER SEQUENCE: 5FAM-GGGPA-Thr(PO3H2)-PKKAKKLMOLECULAR FORMULA: C77H112N17O24P1MOLECULAR WEIGHT:1690.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0293 Supplier CPC Scientific Supplier No. PROK040A
ONE-LETTER SEQUENCE: 5FAM-GGGPA-Thr(PO3H2)-PKKAKKLMOLECULAR FORMULA: C77H112N17O24P1MOLECULAR WEIGHT:1690.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases