Learn More
CPC Scientific 5FAM-Gly-Gly-Gly-Pro-Ala-Thr-Pro-Lys-Lys-Ala-Lys-Lys-Leu-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110295
Encompass
ONE-LETTER SEQUENCE: 5FAM-GGGPATPKKAKKLMOLECULAR FORMULA: C77H111N17O21MOLECULAR WEIGHT:1610.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0295 Supplier CPC Scientific Supplier No. PROK041A
ONE-LETTER SEQUENCE: 5FAM-GGGPATPKKAKKLMOLECULAR FORMULA: C77H111N17O21MOLECULAR WEIGHT:1610.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases