Learn More
CPC Scientific 5FAM-Gly-Gly-Gly-Arg-Ser-Pro-Gly-Arg-Arg-Arg-Arg-Lys-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110269
Encompass
ONE-LETTER SEQUENCE: 5FAM-GGGRSPGRRRRKMOLECULAR FORMULA: C73H108N28O20MOLECULAR WEIGHT:1697.9STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0269 Supplier CPC Scientific Supplier No. PROK027A
ONE-LETTER SEQUENCE: 5FAM-GGGRSPGRRRRKMOLECULAR FORMULA: C73H108N28O20MOLECULAR WEIGHT:1697.9STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases