Learn More
CPC Scientific 5FAM-Gly-Gly-Gly-Arg-Ser-Pro-Gly-Arg-Arg-Arg-Arg-Lys-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110270
Encompass
ONE-LETTER SEQUENCE: 5FAM-GGGRSPGRRRRKMOLECULAR FORMULA: C73H108N28O20MOLECULAR WEIGHT:1697.9STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0270 Supplier CPC Scientific Supplier No. PROK027B
ONE-LETTER SEQUENCE: 5FAM-GGGRSPGRRRRKMOLECULAR FORMULA: C73H108N28O20MOLECULAR WEIGHT:1697.9STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases