Learn More
CPC Scientific 5FAM-Gly-Gly-Leu-Arg-Arg-Ala-Ser-Leu-Gly-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110505
Encompass
ONE-LETTER SEQUENCE: 5FAM-GGLRRASLGMOLECULAR FORMULA: C57H77N15O17MOLECULAR WEIGHT:1244.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0505 Supplier CPC Scientific Supplier No. SUBS032B
ONE-LETTER SEQUENCE: 5FAM-GGLRRASLGMOLECULAR FORMULA: C57H77N15O17MOLECULAR WEIGHT:1244.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases