Learn More
CPC Scientific 5FAM-Gly-Pro-Trp-Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Met-Asp-Phe-Lys-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110297
Encompass
ONE-LETTER SEQUENCE: 5FAM-GPWLEEEEEAYGWMDFK-NH2MOLECULAR FORMULA: C119H141N21O36S1MOLECULAR WEIGHT:2473.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0297 Supplier CPC Scientific Supplier No. PROK042A
ONE-LETTER SEQUENCE: 5FAM-GPWLEEEEEAYGWMDFK-NH2MOLECULAR FORMULA: C119H141N21O36S1MOLECULAR WEIGHT:2473.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases