Learn More
CPC Scientific 5FAM-Gly-Pro-Trp-Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Met-Asp-Phe-Lys-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110298
Encompass
ONE-LETTER SEQUENCE: 5FAM-GPWLEEEEEAYGWMDFK-NH2MOLECULAR FORMULA: C119H141N21O36S1MOLECULAR WEIGHT:2473.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0298 Supplier CPC Scientific Supplier No. PROK042B
ONE-LETTER SEQUENCE: 5FAM-GPWLEEEEEAYGWMDFK-NH2MOLECULAR FORMULA: C119H141N21O36S1MOLECULAR WEIGHT:2473.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases