Learn More
CPC Scientific 5FAM-Gly-Arg-Pro-Arg-Thr-Ser-Ser-Phe-Ala-Glu-Gly-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110497
Encompass
This is a FAM labeled (Abs/Em=492/518 nm) phosphorylated peptide substrate for PP1 and PP2A phosphatases.ONE-LETTER SEQUENCE: 5Fam-GRPRTSSFAEG MOLECULAR FORMULA: C69H87N17O23MOLECULAR WEIGHT:1522.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: Klumpp, M. J. Biomol. Screening 11, 617 (2006)
Catalog No. 50-211-0497 Supplier CPC Scientific Supplier No. SUBS019B
This is a FAM labeled (Abs/Em=492/518 nm) phosphorylated peptide substrate for PP1 and PP2A phosphatases.ONE-LETTER SEQUENCE: 5Fam-GRPRTSSFAEG MOLECULAR FORMULA: C69H87N17O23MOLECULAR WEIGHT:1522.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: Klumpp, M. J. Biomol. Screening 11, 617 (2006)