Learn More
CPC Scientific 5FAM-Gly-Tyr-Ile-Tyr-Gly-Ser-Phe-Lys-Lys-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110242
Encompass
ONE-LETTER SEQUENCE: 5FAM-GYIYGSFKKMOLECULAR FORMULA: C73H85N11O19MOLECULAR WEIGHT:1420.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0242 Supplier CPC Scientific Supplier No. PROK013B
ONE-LETTER SEQUENCE: 5FAM-GYIYGSFKKMOLECULAR FORMULA: C73H85N11O19MOLECULAR WEIGHT:1420.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases