Learn More
CPC Scientific 5FAM-Ile-Arg-Arg-Leu-Ser-Thr-Arg-Arg-Arg-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110235
Encompass
ONE-LETTER SEQUENCE: 5FAM-IRRLSTRRRMOLECULAR FORMULA: C70H106N24O18MOLECULAR WEIGHT:1571.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0235 Supplier CPC Scientific Supplier No. PROK010A
ONE-LETTER SEQUENCE: 5FAM-IRRLSTRRRMOLECULAR FORMULA: C70H106N24O18MOLECULAR WEIGHT:1571.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases