Learn More
CPC Scientific 5FAM-Ile-Arg-Arg-Leu-Ser-Thr-Arg-Arg-Arg-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110236
Encompass
ONE-LETTER SEQUENCE: 5FAM-IRRLSTRRRMOLECULAR FORMULA: C70H106N24O18MOLECULAR WEIGHT:1571.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0236 Supplier CPC Scientific Supplier No. PROK010B
ONE-LETTER SEQUENCE: 5FAM-IRRLSTRRRMOLECULAR FORMULA: C70H106N24O18MOLECULAR WEIGHT:1571.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases