Learn More
CPC Scientific 5FAM-Ile-Tyr-Ser-Gly-Asp-Tyr-Tyr-Arg-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110302
Encompass
SEQUENCE: 5FAM-Ile-Tyr-Ser-Gly-Asp-Tyr-Tyr-Arg-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: 5FAM-IYSGDYYR-NH2MOLECULAR FORMULA: C69H76N12O20MOLECULAR WEIGHT: 1393.4STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES:SKU(s): PROK-044A, PROK-044B
Catalog No. 50-211-0302 Supplier CPC Scientific Supplier No. PROK044B
SEQUENCE: 5FAM-Ile-Tyr-Ser-Gly-Asp-Tyr-Tyr-Arg-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: 5FAM-IYSGDYYR-NH2MOLECULAR FORMULA: C69H76N12O20MOLECULAR WEIGHT: 1393.4STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES:SKU(s): PROK-044A, PROK-044B