Learn More
CPC Scientific 5FAM-Lys-Ala-Ile-Glu-Thr-Asp-Lys-Glu-Tyr-Tyr-Thr-Val-Lys-Asp-NH2 (acetate salt) 1MG
SDP
Catalog No. 502110246
Encompass
ONE-LETTER SEQUENCE: 5FAM-KAIETDKEYYTVKD-NH2MOLECULAR FORMULA: C97H130N18O32MOLECULAR WEIGHT:2060.2STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0246 Supplier CPC Scientific Supplier No. PROK015B
ONE-LETTER SEQUENCE: 5FAM-KAIETDKEYYTVKD-NH2MOLECULAR FORMULA: C97H130N18O32MOLECULAR WEIGHT:2060.2STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases