Learn More
CPC Scientific 5FAM-Lys-Glu-Asp-Pro-Asp-Tyr-Glu-Trp-Pro-Ser-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110243
Encompass
ONE-LETTER SEQUENCE: 5FAM-KEDPDYEWPS-NH2MOLECULAR FORMULA: C78H87N13O26MOLECULAR WEIGHT:1622.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0243 Supplier CPC Scientific Supplier No. PROK014A
ONE-LETTER SEQUENCE: 5FAM-KEDPDYEWPS-NH2MOLECULAR FORMULA: C78H87N13O26MOLECULAR WEIGHT:1622.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases