Learn More
CPC Scientific 5FAM-Lys-Lys-Ala-Leu-His-Arg-Gln-Glu-Thr-Val-Asp-Ala-Leu-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110307
Encompass
ONE-LETTER SEQUENCE: 5FAM-KKALHRQETVDAL-NH2MOLECULAR FORMULA: C86H124N22O25MOLECULAR WEIGHT:1866.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0307 Supplier CPC Scientific Supplier No. PROK047A
ONE-LETTER SEQUENCE: 5FAM-KKALHRQETVDAL-NH2MOLECULAR FORMULA: C86H124N22O25MOLECULAR WEIGHT:1866.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases