Learn More
CPC Scientific 5FAM-Lys-Lys-Lys-Lys-Lys-Arg-Phe-Ser-Phe-Lys-Lys-Ala-Phe-Lys-Leu-Ala-Gly-Phe-Ala-Phe-Lys-Lys-Asn-Lys-Lys-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110274
Encompass
ONE-LETTER SEQUENCE: 5FAM-KKKKKRFSFKKAFKLAGFAFKKNKKMOLECULAR FORMULA: C168H253N41O34MOLECULAR WEIGHT:3391.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0274 Supplier CPC Scientific Supplier No. PROK030B
ONE-LETTER SEQUENCE: 5FAM-KKKKKRFSFKKAFKLAGFAFKKNKKMOLECULAR FORMULA: C168H253N41O34MOLECULAR WEIGHT:3391.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases