Learn More
CPC Scientific 5FAM-Lys-Lys-Leu-Ala-Thr-Gly-Asp-Tyr-Met-Asn-Met-Ser-Pro-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110258
Encompass
ONE-LETTER SEQUENCE: 5FAM-KKLATGDYMNMSP-NH2MOLECULAR FORMULA: C83H113N17O25S2MOLECULAR WEIGHT:1813.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0258 Supplier CPC Scientific Supplier No. PROK021B
ONE-LETTER SEQUENCE: 5FAM-KKLATGDYMNMSP-NH2MOLECULAR FORMULA: C83H113N17O25S2MOLECULAR WEIGHT:1813.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases