Learn More
CPC Scientific 5FAM-Lys-Lys-Arg-Lys-Ser-Ser-Leu-Arg-Arg-Trp-Ser-Pro-Leu-Thr-Pro-Arg-Gln-Met-Ser-Phe-Asp-Cys-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110260
Encompass
ONE-LETTER SEQUENCE: 5FAM-KKRKSSLRRWSPLTPRQMSFDC-NH2MOLECULAR FORMULA: C138H206N40O36S2MOLECULAR WEIGHT:3065.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0260 Supplier CPC Scientific Supplier No. PROK022B
ONE-LETTER SEQUENCE: 5FAM-KKRKSSLRRWSPLTPRQMSFDC-NH2MOLECULAR FORMULA: C138H206N40O36S2MOLECULAR WEIGHT:3065.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases