Learn More
CPC Scientific 5FAM-Lys-Lys-Arg-Asn-Arg-Arg-Leu-Ser-Val-Ala-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110310
Encompass
ONE-LETTER SEQUENCE: 5FAM-KKRNRRLSVAMOLECULAR FORMULA: C72H108N22O19MOLECULAR WEIGHT:1585.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0310 Supplier CPC Scientific Supplier No. PROK048B
ONE-LETTER SEQUENCE: 5FAM-KKRNRRLSVAMOLECULAR FORMULA: C72H108N22O19MOLECULAR WEIGHT:1585.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases