Learn More
CPC Scientific 5FAM-Lys-Lys-Arg-Asn-Arg-Thr-Leu-Thr-Lys-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110252
Encompass
ONE-LETTER SEQUENCE: 5FAM-KKRNRTLTKMOLECULAR FORMULA: C69H103N19O19MOLECULAR WEIGHT:1502.7STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0252 Supplier CPC Scientific Supplier No. PROK018B
ONE-LETTER SEQUENCE: 5FAM-KKRNRTLTKMOLECULAR FORMULA: C69H103N19O19MOLECULAR WEIGHT:1502.7STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases