Learn More
CPC Scientific 5FAM-Lys-Lys-Arg-Pro-Gln-Arg-Arg-Tyr-Ser-Asn-Val-Phe-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110311
Encompass
ONE-LETTER SEQUENCE: 5FAM-KKRPQRRYSNVFMOLECULAR FORMULA: C91H125N25O23MOLECULAR WEIGHT:1937.2STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0311 Supplier CPC Scientific Supplier No. PROK049A
ONE-LETTER SEQUENCE: 5FAM-KKRPQRRYSNVFMOLECULAR FORMULA: C91H125N25O23MOLECULAR WEIGHT:1937.2STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases