Learn More
CPC Scientific 5FAM-Lys-Arg-Arg-Glu-Ile-Leu-Ser-Arg-Arg-Pro-Ser-Tyr-Arg-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110278
Encompass
ONE-LETTER SEQUENCE: 5FAM-KRREILSRRPSYR-NH2MOLECULAR FORMULA: C94H140N30O24MOLECULAR WEIGHT:2074.4STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0278 Supplier CPC Scientific Supplier No. PROK032B
ONE-LETTER SEQUENCE: 5FAM-KRREILSRRPSYR-NH2MOLECULAR FORMULA: C94H140N30O24MOLECULAR WEIGHT:2074.4STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases