Learn More
CPC Scientific 5FAM-Leu-Lys-Lys-Leu-Arg-Arg-Arg-Leu-Ser-Asp-Ala-Asn-Phe-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110275
Encompass
ONE-LETTER SEQUENCE: 5FAM-LKKLRRRLSDANF-NH2MOLECULAR FORMULA: C92H136N26O23MOLECULAR WEIGHT:1974.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0275 Supplier CPC Scientific Supplier No. PROK031A
ONE-LETTER SEQUENCE: 5FAM-LKKLRRRLSDANF-NH2MOLECULAR FORMULA: C92H136N26O23MOLECULAR WEIGHT:1974.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases