Learn More
CPC Scientific 5FAM-Leu-Lys-Lys-Leu-Thr-Arg-Arg-Ala-Ser-Phe-Ser-Gly-Gln-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110304
Encompass
ONE-LETTER SEQUENCE: 5FAM-LKKLTRRASFSGQMOLECULAR FORMULA: C86H124N22O24MOLECULAR WEIGHT:1850.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0304 Supplier CPC Scientific Supplier No. PROK045B
ONE-LETTER SEQUENCE: 5FAM-LKKLTRRASFSGQMOLECULAR FORMULA: C86H124N22O24MOLECULAR WEIGHT:1850.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases