Learn More
CPC Scientific 5FAM-Met-His-Arg-Gln-Glu-Thr-Val-Asp-Cys-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110508
Encompass
ONE-LETTER SEQUENCE: 5FAM-MHRQETVDC-NH2MOLECULAR FORMULA: C64H82N16O21S2MOLECULAR WEIGHT:1475.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0508 Supplier CPC Scientific Supplier No. SUBS034A
ONE-LETTER SEQUENCE: 5FAM-MHRQETVDC-NH2MOLECULAR FORMULA: C64H82N16O21S2MOLECULAR WEIGHT:1475.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases