Learn More
CPC Scientific 5FAM-Asn-Val-Lys-Ser-Lys-Ile-Gly-Ser-Thr-Glu-Asn-Leu-Lys-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110510
Encompass
ONE-LETTER SEQUENCE: 5FAM-NVKSKIGSTENLK-NH2MOLECULAR FORMULA: C81H119N19O26MOLECULAR WEIGHT:1775STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0510 Supplier CPC Scientific Supplier No. SUBS035A
ONE-LETTER SEQUENCE: 5FAM-NVKSKIGSTENLK-NH2MOLECULAR FORMULA: C81H119N19O26MOLECULAR WEIGHT:1775STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases