Learn More
CPC Scientific 5FAM-Pro-Lys-Thr-Pro-Lys-Lys-Ala-Lys-Lys-Leu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PROK019B
Encompass
ONE-LETTER SEQUENCE: 5FAM-PKTPKKAKKLMOLECULAR FORMULA: C74H109N15O18MOLECULAR WEIGHT:1496.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0254