Learn More
CPC Scientific 5FAM-Pro-Leu-Arg-Arg-Thr-Leu-Ser-Val-Ala-Ala-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110514
Encompass
ONE-LETTER SEQUENCE: 5FAM-PLRRTLSVAAMOLECULAR FORMULA: C68H96N16O19MOLECULAR WEIGHT:1441.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0514 Supplier CPC Scientific Supplier No. SUBS037A
ONE-LETTER SEQUENCE: 5FAM-PLRRTLSVAAMOLECULAR FORMULA: C68H96N16O19MOLECULAR WEIGHT:1441.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases