Learn More
CPC Scientific 5FAM-Gln-Asn-Pro-Ser-Arg-Cys-Ser-Val-Ser-Leu-Ser-Asn-Val-Glu-Ala-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110517
Encompass
ONE-LETTER SEQUENCE: 5FAM-QNPSRCSVSLSNVEA-NH2MOLECULAR FORMULA: C84H118N22O30S1MOLECULAR WEIGHT:1948.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0517 Supplier CPC Scientific Supplier No. SUBS038B
ONE-LETTER SEQUENCE: 5FAM-QNPSRCSVSLSNVEA-NH2MOLECULAR FORMULA: C84H118N22O30S1MOLECULAR WEIGHT:1948.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases