Learn More
CPC Scientific 5FAM-Arg-Lys-Arg-Ser-Arg-Ala-Glu-Ala-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110519
Encompass
ONE-LETTER SEQUENCE: 5FAM-RKRSRAEA-NH2MOLECULAR FORMULA: C59H83N19O17MOLECULAR WEIGHT:1330.4STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0519 Supplier CPC Scientific Supplier No. SUBS039B
ONE-LETTER SEQUENCE: 5FAM-RKRSRAEA-NH2MOLECULAR FORMULA: C59H83N19O17MOLECULAR WEIGHT:1330.4STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases