Learn More
CPC Scientific 5FAM-Arg-Pro-Arg-Thr-Ser-Ser-Phe-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110238
Encompass
SEQUENCE: 5FAM-Arg-Pro-Arg-Thr-Ser-Ser-Phe-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: 5FAM-RPRTSSFMOLECULAR FORMULA: C57H69N13O17MOLECULAR WEIGHT: 1208.3STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES:SKU(s): PROK-011A, PROK-011B
Catalog No. 50-211-0238 Supplier CPC Scientific Supplier No. PROK011B
SEQUENCE: 5FAM-Arg-Pro-Arg-Thr-Ser-Ser-Phe-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: 5FAM-RPRTSSFMOLECULAR FORMULA: C57H69N13O17MOLECULAR WEIGHT: 1208.3STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES:SKU(s): PROK-011A, PROK-011B