Learn More
CPC Scientific 5FAM-Arg-Gln-Met-Ser-Phe-Arg-Leu-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110264
Encompass
SEQUENCE: 5FAM-Arg-Gln-Met-Ser-Phe-Arg-Leu-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: 5FAM-RQMSFRLMOLECULAR FORMULA: C61H78N14O16S1MOLECULAR WEIGHT: 1295.5STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES:SKU(s): PROK-024A, PROK-024B
Catalog No. 50-211-0264 Supplier CPC Scientific Supplier No. PROK024B
SEQUENCE: 5FAM-Arg-Gln-Met-Ser-Phe-Arg-Leu-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: 5FAM-RQMSFRLMOLECULAR FORMULA: C61H78N14O16S1MOLECULAR WEIGHT: 1295.5STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES:SKU(s): PROK-024A, PROK-024B