Learn More
CPC Scientific 5FAM-Arg-Arg-Phe-Arg-Phe-Thr-Ala-Asn-Leu-Ser-Lys-Arg-Tyr-Glu-Ala-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110521
Encompass
ONE-LETTER SEQUENCE: 5FAM-RRFRFTANLSKRYEA-NH2MOLECULAR FORMULA: C106H146N30O27MOLECULAR WEIGHT:2272.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0521 Supplier CPC Scientific Supplier No. SUBS040B
ONE-LETTER SEQUENCE: 5FAM-RRFRFTANLSKRYEA-NH2MOLECULAR FORMULA: C106H146N30O27MOLECULAR WEIGHT:2272.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases