Learn More
CPC Scientific 5FAM-Thr-Glu-Ser-Gln-Ser-Leu-Thr-Leu-Thr-Asp-Val-Glu-Asn-Leu-His-Leu-Pro-Leu-Pro-Leu-Leu-Gln-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110265
Encompass
ONE-LETTER SEQUENCE: 5FAM-TESQSLTLTDVENLHLPLPLLQ-NH2MOLECULAR FORMULA: C130H192N28O42MOLECULAR WEIGHT:2819.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0265 Supplier CPC Scientific Supplier No. PROK025A
ONE-LETTER SEQUENCE: 5FAM-TESQSLTLTDVENLHLPLPLLQ-NH2MOLECULAR FORMULA: C130H192N28O42MOLECULAR WEIGHT:2819.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases