Learn More
Cayman Chemical 5FAMLPETGGtrIfluoroACTAt 10mg
SDP
Catalog No. 502915471
Encompass_Preferred
A fluorescent peptide; ex/em = 492/518 nm, respectively; has been used for site-specific labeling of target proteins via sortase-catalyzed transpeptidation
Catalog No. 50-291-5471 Supplier Cayman Chemical Supplier No. 3699710 MG
A fluorescent peptide; ex/em = 492/518 nm, respectively; has been used for site-specific labeling of target proteins via sortase-catalyzed transpeptidation